The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138423.3 | NP_612432.2 | 5 - 6 | 1040 | 71 |
NM_177974.2 | NP_816929.1 | 5 - 6 | 1040 | 71 | |
XM_017021880.1 | XP_016877369.1 | 5 - 6 | 1135 | 71 | |
GenBank mRNA | AK095068.1 | - | 1 - 2 | 80 | 71 |
AK291026.1 | - | 5 - 6 | 797 | 71 | |
AK298564.1 | - | 6 - 7 | 755 | 71 | |
AL832333.1 | - | 5 - 6 | 812 | 71 | |
AL832840.1 | - | 5 - 6 | 482 | 71 | |
AY358086.1 | - | 5 - 6 | 991 | 71 | |
BC012124.1 | - | 4 - 5 | 929 | 71 | |
BC063480.1 | - | 5 - 6 | 994 | 71 |