Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21B
Entrez Gene ID
112885
Gene Name
PHD finger protein 21B
Gene Aliases
BHC80L, PHF4
Chromosome Location
Chr.22: 44881162 - 45009928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135862.2 NP_001129334.1 2 - 3 162 62
NM_138415.4 NP_612424.1 1 - 2 207 62
NR_134535.1 1 - 2 117 62
XM_006724122.1 XP_006724185.1 1 - 2 342 62
XM_011529855.1 XP_011528157.1 1 - 2 342 62
GenBank mRNA AK092243.1 - 2 - 3 162 62
BC012187.1 - 1 - 2 207 62
CR456448.1 - 1 - 2 89 62
KU178842.1 - 1 - 2 56 62
KU178843.1 - 1 - 2 56 62
KU178844.1 - 1 - 2 56 62

View Full Product Details