Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR31
Entrez Gene ID
114987
Gene Name
WD repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.9: 113313222 - 113340340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006615.2 NP_001006616.1 7 - 8 805 99
NM_001012361.3 NP_001012361.1 8 - 9 938 99
NM_145241.4 NP_660284.1 8 - 9 935 99
XM_011518194.2 XP_011516496.1 7 - 8 849 99
GenBank mRNA AK074891.1 - 7 - 8 719 99
AK093240.1 - 8 - 9 867 99
AK313843.1 - 8 - 9 877 99
BC012352.1 - 8 - 9 867 99
BC022982.1 - 8 - 9 864 99

View Full Product Details