The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164796.1 | NP_001158268.1 | 8 - 9 | 904 | 60 |
NM_173471.3 | NP_775742.4 | 10 - 11 | 1436 | 60 | |
NR_028475.1 | 9 - 10 | 989 | 60 | ||
XM_006712956.1 | XP_006713019.1 | 8 - 9 | 972 | 60 | |
XM_011533327.2 | XP_011531629.1 | 9 - 10 | 799 | 60 | |
XM_017005671.1 | XP_016861160.1 | 10 - 11 | 1053 | 60 | |
XM_017005672.1 | XP_016861161.1 | 10 - 11 | 835 | 60 | |
XM_017005673.1 | XP_016861162.1 | 9 - 10 | 657 | 60 | |
XM_017005674.1 | XP_016861163.1 | 7 - 8 | 611 | 60 | |
GenBank mRNA | AJ580932.1 | - | 9 - 10 | 783 | 60 |
AK092495.1 | - | 10 - 11 | 1436 | 60 | |
AK096876.1 | - | 9 - 10 | 793 | 60 | |
AK291873.1 | - | 9 - 10 | 634 | 60 | |
AX747571.1 | - | 10 - 11 | 1436 | 60 | |
BC003399.1 | - | 9 - 10 | 683 | 60 | |
BC012852.1 | - | 8 - 9 | 623 | 60 | |
CR749718.1 | - | 9 - 10 | 636 | 60 |