Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTI1
Entrez Gene ID
9675
Gene Name
TELO2 interacting protein 1
Gene Aliases
KIAA0406, smg-10
Chromosome Location
Chr.20: 37983007 - 38033468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303457.1 NP_001290386.1 5 - 6 2899 98
NM_014657.2 NP_055472.1 6 - 7 3039 98
XM_017028148.1 XP_016883637.1 5 - 6 2899 98
GenBank mRNA AB007866.2 - 5 - 6 2988 98
AK308164.1 - 5 - 6 2888 98
BC013121.1 - 6 - 7 3002 98
BC013755.1 - 5 - 6 2852 98
BC131759.1 - 4 - 5 574 98

View Full Product Details