Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP2
Entrez Gene ID
26157
Gene Name
GTPase, IMAP family member 2
Gene Aliases
HIMAP2, IAN12, IMAP2
Chromosome Location
Chr.7: 150685706 - 150693641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015660.2 NP_056475.1 2 - 3 116 61
GenBank mRNA AK074199.1 - 2 - 3 115 61
AL110151.1 - 2 - 3 95 61
BC013934.1 - 2 - 3 116 61
BC032345.1 - 2 - 3 92 61
BG687383.1 - 2 - 3 117 61

View Full Product Details