The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270940.1 | NP_001257869.1 | 20 - 21 | 3296 | 75 |
NM_015171.3 | NP_055986.1 | 19 - 20 | 3160 | 75 | |
XM_005255197.3 | XP_005255254.1 | 17 - 18 | 2494 | 75 | |
XM_005255198.1 | XP_005255255.1 | 15 - 16 | 2643 | 75 | |
XM_005255199.2 | XP_005255256.1 | 15 - 16 | 1837 | 75 | |
GenBank mRNA | AB002368.3 | - | 19 - 20 | 3111 | 75 |
AK026172.1 | - | 16 - 17 | 2304 | 75 | |
AK123846.1 | - | 20 - 21 | 2928 | 75 | |
AK223043.1 | - | 3 - 4 | 470 | 75 | |
AY026388.1 | - | 19 - 20 | 2633 | 75 | |
BC004403.1 | - | 13 - 14 | 1900 | 75 | |
BC014071.2 | - | 13 - 14 | 1900 | 75 | |
BC078674.1 | - | 15 - 16 | 2105 | 75 | |
BC108286.1 | - | 15 - 16 | 2059 | 75 | |
BC130304.1 | - | 19 - 20 | 2633 | 75 | |
BC144189.1 | - | 19 - 20 | 2633 | 75 |