The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138785.3 | NP_620140.1 | 4 - 5 | 452 | 129 |
GenBank mRNA | AK289402.1 | - | 4 - 5 | 439 | 129 |
AK295690.1 | - | 3 - 4 | 381 | 129 | |
AK311061.1 | - | 2 - 3 | 279 | 129 | |
AK315729.1 | - | 4 - 5 | 449 | 129 | |
AY358952.1 | - | 4 - 5 | 570 | 129 | |
BC014320.1 | - | 4 - 5 | 456 | 129 |