The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 62 - 63 | 9951 | 91 |
XM_005262858.4 | XP_005262915.1 | 64 - 65 | 9992 | 91 | |
XM_011531757.2 | XP_011530059.1 | 63 - 64 | 9903 | 91 | |
XM_011531759.2 | XP_011530061.1 | 62 - 63 | 9865 | 91 | |
XM_011531760.2 | XP_011530062.1 | 62 - 63 | 9803 | 91 | |
XM_011531761.2 | XP_011530063.1 | 63 - 64 | 9858 | 91 | |
XM_011531762.2 | XP_011530064.1 | 62 - 63 | 9852 | 91 | |
XM_011531763.2 | XP_011530065.1 | 61 - 62 | 9749 | 91 | |
XM_011531764.2 | XP_011530066.1 | 62 - 63 | 9819 | 91 | |
XM_011531765.2 | XP_011530067.1 | 59 - 60 | 9554 | 91 | |
XM_017007906.1 | XP_016863395.1 | 65 - 66 | 10120 | 91 | |
XM_017007907.1 | XP_016863396.1 | 65 - 66 | 10117 | 91 | |
XM_017007908.1 | XP_016863397.1 | 64 - 65 | 10049 | 91 | |
XM_017007909.1 | XP_016863398.1 | 50 - 51 | 7745 | 91 | |
GenBank mRNA | AB023210.2 | - | 27 - 28 | 3634 | 91 |
AF538685.1 | - | 61 - 62 | 9843 | 91 | |
AK055806.1 | - | 13 - 14 | 1598 | 91 | |
AK092394.1 | - | 2 - 3 | 330 | 91 | |
BC015214.1 | - | 2 - 3 | 277 | 91 | |
BC065502.1 | - | 9 - 10 | 1301 | 91 | |
BC150246.1 | - | 27 - 28 | 3655 | 91 | |
BX648077.1 | - | 19 - 20 | 2516 | 91 | |
BX648153.1 | - | 22 - 23 | 2844 | 91 |