The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114176.1 | NP_001107648.1 | 2 - 3 | 900 | 107 |
NM_001267043.1 | NP_001253972.1 | 2 - 3 | 350 | 107 | |
NM_153207.4 | NP_694939.2 | 2 - 3 | 900 | 107 | |
XM_006719031.3 | XP_006719094.1 | 2 - 3 | 1082 | 107 | |
XM_017018806.1 | XP_016874295.1 | 2 - 3 | 227 | 107 | |
GenBank mRNA | AB209384.1 | - | 2 - 3 | 694 | 107 |
AK126973.1 | - | 3 - 4 | 686 | 107 | |
AK131361.1 | - | 2 - 3 | 900 | 107 | |
AK131410.1 | - | 2 - 3 | 350 | 107 | |
BC015624.2 | - | 2 - 3 | 387 | 107 | |
BC022220.1 | - | 2 - 3 | 875 | 107 |