The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173575.3 | NP_775846.2 | 1 - 2 | 403 | 62 |
XM_011539690.2 | XP_011537992.1 | 1 - 2 | 374 | 62 | |
GenBank mRNA | AY098866.1 | - | 1 - 2 | 403 | 62 |