The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032437.3 | NP_115813.2 | 1 - 2 | 333 | 97 |
XM_005271274.3 | XP_005271331.1 | 1 - 2 | 133 | 97 | |
GenBank mRNA | AB058702.1 | - | 1 - 2 | 96 | 97 |
AK092219.1 | - | 1 - 2 | 244 | 97 | |
AK292220.1 | - | 1 - 2 | 292 | 97 | |
AK315850.1 | - | 1 - 2 | 109 | 97 | |
BC015814.1 | - | 1 - 2 | 37 | 97 | |
HY084741.1 | - | 1 - 2 | 333 | 97 |