Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr.15: 42158701 - 42208331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301138.1 NP_001288067.1 11 - 12 1167 76
NM_015289.3 NP_056104.2 10 - 11 1134 76
XM_011521403.2 XP_011519705.1 11 - 12 1136 76
XM_011521404.2 XP_011519706.1 10 - 11 1139 76
XM_017022036.1 XP_016877525.1 11 - 12 1135 76
XM_017022037.1 XP_016877526.1 10 - 11 1139 76
XM_017022038.1 XP_016877527.1 10 - 11 1322 76
XM_017022039.1 XP_016877528.1 10 - 11 1107 76
GenBank mRNA AB018313.2 - 10 - 11 1085 76
AF280814.1 - 11 - 12 994 76
AF281052.1 - 10 - 11 1104 76
BC015817.2 - 6 - 7 694 76
BC068559.1 - 10 - 11 1103 76
BX537404.1 - 10 - 11 1113 76

View Full Product Details