The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145418.1 | NP_001138890.1 | 10 - 11 | 3690 | 65 |
XM_005261405.2 | XP_005261462.1 | 10 - 11 | 3690 | 65 | |
XM_006724171.3 | XP_006724234.1 | 9 - 10 | 12468 | 65 | |
XM_011530018.2 | XP_011528320.1 | 10 - 11 | 3869 | 65 | |
XM_011530019.2 | XP_011528321.1 | 10 - 11 | 3690 | 65 | |
XM_011530020.1 | XP_011528322.1 | 10 - 11 | 3690 | 65 | |
XM_011530021.2 | XP_011528323.1 | 10 - 11 | 3690 | 65 | |
XM_011530022.1 | XP_011528324.1 | 10 - 11 | 3690 | 65 | |
XM_017028673.1 | XP_016884162.1 | 10 - 11 | 3663 | 65 | |
GenBank mRNA | AB028966.3 | - | 5 - 6 | 2483 | 65 |
AB665284.1 | - | 10 - 11 | 3638 | 65 | |
BC016465.1 | - | 2 - 3 | 234 | 65 |