Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
Chromosome Location
Chr.17: 43527843 - 43546432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079675.2 NP_001073143.1 6 - 7 683 61
NM_001261437.1 NP_001248366.1 5 - 6 572 61
NM_001261438.1 NP_001248367.1 5 - 6 401 61
NM_001986.2 NP_001977.1 6 - 7 512 61
XM_011524514.1 XP_011522816.1 4 - 5 324 61
XM_017024349.1 XP_016879838.1 6 - 7 474 61
GenBank mRNA AK290429.1 - 6 - 7 474 61
AK312934.1 - 6 - 7 471 61
AK315961.1 - 5 - 6 463 61
BC016623.1 - 6 - 7 454 61
BM013535.1 - 6 - 7 512 61
D12765.1 - 4 - 5 318 61
DQ396625.1 - 4 - 5 391 61
DQ396626.1 - 4 - 5 357 61
EU693078.1 - - 310 61
U18018.1 - 6 - 7 587 61

View Full Product Details