The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032430.1 | NP_115806.1 | 16 - 17 | 2166 | 82 |
XM_005259327.3 | XP_005259384.1 | 16 - 17 | 2235 | 82 | |
XM_011527395.2 | XP_011525697.2 | 14 - 15 | 1600 | 82 | |
GenBank mRNA | AB058714.1 | - | 15 - 16 | 1701 | 82 |
AF479826.1 | - | 18 - 19 | 2064 | 82 | |
AF479827.1 | - | 16 - 17 | 2166 | 82 | |
AK124999.1 | - | 8 - 9 | 1135 | 82 | |
AL831945.1 | - | 1 - 2 | 93 | 82 | |
AL834275.1 | - | 16 - 17 | 1789 | 82 | |
AY458602.1 | - | 16 - 17 | 1938 | 82 | |
AY505124.1 | - | 16 - 17 | 1889 | 82 | |
BC016681.1 | - | 8 - 9 | 1200 | 82 |