The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017969.2 | NP_060439.2 | 13 - 14 | 2631 | 70 |
XM_005263707.3 | XP_005263764.1 | 13 - 14 | 2467 | 70 | |
XM_017004468.1 | XP_016859957.1 | 13 - 14 | 2401 | 70 | |
GenBank mRNA | AK001717.1 | - | 8 - 9 | 876 | 70 |
AK056881.1 | - | 13 - 14 | 1632 | 70 | |
AK294669.1 | - | 14 - 15 | 2339 | 70 | |
AK296848.1 | - | 14 - 15 | 2439 | 70 | |
AL834178.1 | - | 13 - 14 | 2579 | 70 | |
BC017012.2 | - | 10 - 11 | 1108 | 70 | |
BC110536.1 | - | 13 - 14 | 2461 | 70 | |
BC110537.1 | - | 13 - 14 | 2461 | 70 |