The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000878.3 | NP_000869.1 | 3 - 4 | 337 | 76 |
GenBank mRNA | AK312860.1 | - | 3 - 4 | 331 | 76 |
BC025691.1 | - | 3 - 4 | 320 | 76 | |
CR456506.1 | - | 2 - 3 | 225 | 76 | |
M26062.1 | - | 3 - 4 | 337 | 76 |