The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001041.3 | NP_001032.2 | 8 - 9 | 968 | 103 |
XM_011513078.2 | XP_011511380.1 | 8 - 9 | 972 | 103 | |
GenBank mRNA | BC115034.1 | - | 8 - 9 | 981 | 103 |
BC116452.1 | - | 8 - 9 | 103 | ||
BC116453.1 | - | 8 - 9 | 981 | 103 | |
BC132834.1 | - | 8 - 9 | 961 | 103 | |
BC132860.1 | - | 8 - 9 | 961 | 103 | |
M22616.1 | - | 7 - 8 | 910 | 103 | |
X63597.1 | - | 8 - 9 | 967 | 103 |