The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001041.3 | NP_001032.2 | 14 - 15 | 1659 | 139 |
XM_011513078.2 | XP_011511380.1 | 14 - 15 | 1663 | 139 | |
GenBank mRNA | BC115034.1 | - | 14 - 15 | 1672 | 139 |
BC116453.1 | - | 14 - 15 | 1672 | 139 | |
BC132834.1 | - | 14 - 15 | 1652 | 139 | |
BC132860.1 | - | 14 - 15 | 1652 | 139 | |
M22616.1 | - | 13 - 14 | 1601 | 139 | |
X63597.1 | - | 14 - 15 | 1658 | 139 |