Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO5
Entrez Gene ID
2330
Gene Name
flavin containing monooxygenase 5
Gene Aliases
-
Chromosome Location
Chr.1: 147183963 - 147225798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144829.2 NP_001138301.1 5 - 6 1028 139
NM_001144830.2 NP_001138302.1 4 - 5 1238 139
NM_001461.3 NP_001452.2 5 - 6 1018 139
XM_005272946.4 XP_005273003.1 5 - 6 992 139
XM_005272947.4 XP_005273004.1 5 - 6 736 139
XM_005272948.4 XP_005273005.1 5 - 6 719 139
XM_006711244.3 XP_006711307.1 5 - 6 1008 139
XM_006711245.3 XP_006711308.1 5 - 6 1008 139
XM_011509350.2 XP_011507652.1 5 - 6 1023 139
XM_011509351.2 XP_011507653.1 4 - 5 755 139
XM_017000801.1 XP_016856290.1 4 - 5 767 139
XM_017000802.1 XP_016856291.1 4 - 5 537 139
GenBank mRNA AK022172.1 - 4 - 5 1228 139
AK222728.1 - 5 - 6 733 139
AK225739.1 - 5 - 6 743 139
AK314647.1 - 5 - 6 946 139
BC035687.1 - 4 - 5 911 139
L37080.1 - 5 - 6 704 139
Z47553.1 - 5 - 6 710 139

View Full Product Details