The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166449.1 | NP_001159921.1 | 21 - 22 | 2694 | 64 |
NM_002218.4 | NP_002209.2 | 23 - 24 | 2784 | 64 | |
GenBank mRNA | AB209388.1 | - | 16 - 17 | 4930 | 64 |
AK298407.1 | - | 20 - 21 | 2629 | 64 | |
AK298412.1 | - | 21 - 22 | 2665 | 64 | |
BC136392.1 | - | 23 - 24 | 2771 | 64 | |
BC136393.1 | - | 23 - 24 | 2756 | 64 | |
BX647322.1 | - | 17 - 18 | 2090 | 64 | |
D38595.1 | - | 23 - 24 | 2758 | 64 |