Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53431720-53432228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
UniGene
Hs.171280
Gene Chromosome Location
Chr.X: 53431258-53434376 on Build GRCh38
Transcripts
NM_001037811.2 NM_004493.2

SNP Detail

SNP ID Chromosome Location
rs782167456 Chr.X: 53431928-53431928 on Build GRCh38
rs122462164 Chr.X: 53431819-53431819 on Build GRCh38
rs782612713 Chr.X: 53431792-53431792 on Build GRCh38
rs374264193 Chr.X: 53431912-53431912 on Build GRCh38
rs782240056 Chr.X: 53432130-53432130 on Build GRCh38
rs797045616 Chr.X: 53432108-53432108 on Build GRCh38
rs28935476 Chr.X: 53432110-53432110 on Build GRCh38
rs369860435 Chr.X: 53431747-53431747 on Build GRCh38
rs201631203 Chr.X: 53432158-53432158 on Build GRCh38
rs782388358 Chr.X: 53431862-53431862 on Build GRCh38
rs112756915 Chr.X: 53432119-53432119 on Build GRCh38
rs28935475 Chr.X: 53432086-53432086 on Build GRCh38
rs897659814 Chr.X: 53431791-53431791 on Build GRCh38
rs949464841 Chr.X: 53431985-53431985 on Build GRCh38
rs782464134 Chr.X: 53432043-53432043 on Build GRCh38
rs370838884 Chr.X: 53431963-53431963 on Build GRCh38
rs782209343 Chr.X: 53431843-53431843 on Build GRCh38
rs782565608 Chr.X: 53432045-53432045 on Build GRCh38
rs190913381 Chr.X: 53431979-53431979 on Build GRCh38
rs781883925 Chr.X: 53432030-53432030 on Build GRCh38
rs782407792 Chr.X: 53431937-53431937 on Build GRCh38
rs782672728 Chr.X: 53431768-53431768 on Build GRCh38
rs370477229 Chr.X: 53432114-53432114 on Build GRCh38
rs782644415 Chr.X: 53432129-53432129 on Build GRCh38
rs886037927 Chr.X: 53431801-53431801 on Build GRCh38
rs1036579268 Chr.X: 53431776-53431776 on Build GRCh38
rs781827718 Chr.X: 53431976-53431976 on Build GRCh38
rs782212120 Chr.X: 53432182-53432182 on Build GRCh38
rs782282260 Chr.X: 53431753-53431753 on Build GRCh38
rs373822420 Chr.X: 53431973-53431973 on Build GRCh38
rs782309721 Chr.X: 53432057-53432057 on Build GRCh38
rs1009827132 Chr.X: 53432186-53432186 on Build GRCh38
rs377646597 Chr.X: 53431935-53431935 on Build GRCh38
rs782309408 Chr.X: 53431778-53431778 on Build GRCh38
rs894038747 Chr.X: 53432183-53432183 on Build GRCh38
rs187015851 Chr.X: 53432170-53432170 on Build GRCh38
rs369677190 Chr.X: 53432178-53432178 on Build GRCh38
rs905186398 Chr.X: 53432169-53432169 on Build GRCh38
rs782197443 Chr.X: 53431770-53431770 on Build GRCh38
rs782609456 Chr.X: 53431919-53431919 on Build GRCh38
rs782680843 Chr.X: 53432162-53432162 on Build GRCh38
rs182999706 Chr.X: 53432021-53432021 on Build GRCh38

View Full Product Details