Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDGFRB
Entrez Gene ID
5159
Gene Name
platelet derived growth factor receptor beta
Gene Aliases
CD140B, IBGC4, IMF1, JTK12, KOGS, PDGFR, PDGFR-1, PDGFR1, PENTT
Chromosome Location
Chr.5: 150113839 - 150155884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002609.3 NP_002600.1 16 - 17 2813 79
XM_005268464.2 XP_005268521.1 15 - 16 2792 79
XM_011537658.1 XP_011535960.1 16 - 17 2388 79
XM_011537659.1 XP_011535961.1 17 - 18 3293 79
GenBank mRNA AB209657.1 - 16 - 17 2846 79
AK293093.1 - 16 - 17 2649 79
BC032224.1 - 16 - 17 2819 79
J03278.1 - 16 - 17 2700 79
M21616.1 - 16 - 17 2530 79

View Full Product Details