The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004959.4 | NP_004950.2 | 5 - 6 | 1176 | 67 |
XM_005251871.3 | XP_005251928.1 | 5 - 6 | 1204 | 67 | |
XM_011518455.2 | XP_011516757.1 | 5 - 6 | 1054 | 67 | |
GenBank mRNA | AB307718.1 | - | 4 - 5 | 989 | 67 |
BC032501.1 | - | 5 - 6 | 1176 | 67 | |
HQ692856.1 | - | 4 - 5 | 989 | 67 | |
U76388.1 | - | 4 - 5 | 1004 | 67 |