Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 17 - 18 3416 64
XM_006718397.2 XP_006718460.1 22 - 23 4354 64
XM_006718398.3 XP_006718461.1 18 - 19 4966 64
XM_011520504.1 XP_011518806.1 23 - 24 4397 64
XM_011520507.2 XP_011518809.1 22 - 23 4318 64
XM_011520508.1 XP_011518810.1 22 - 23 4354 64
XM_011520510.1 XP_011518812.1 22 - 23 4270 64
XM_011520511.1 XP_011518813.1 22 - 23 4243 64
XM_011520512.1 XP_011518814.1 18 - 19 3618 64
XM_011520513.1 XP_011518815.1 18 - 19 3616 64
XM_011520514.2 XP_011518816.1 18 - 19 3572 64
XM_011520515.1 XP_011518817.1 19 - 20 3785 64
XM_011520516.1 XP_011518818.1 19 - 20 3785 64
XM_011520517.2 XP_011518819.1 19 - 20 5107 64
XM_011520518.1 XP_011518820.1 19 - 20 3755 64
XM_011520519.1 XP_011518821.1 18 - 19 3643 64
XM_011520522.2 XP_011518824.1 17 - 18 4821 64
XM_011520523.2 XP_011518825.1 18 - 19 5703 64
XM_011520526.2 XP_011518828.1 10 - 11 2672 64
XM_011520527.1 XP_011518829.1 10 - 11 1940 64
XM_017018629.1 XP_016874118.1 22 - 23 4747 64
XM_017018630.1 XP_016874119.1 17 - 18 3709 64
XM_017018631.1 XP_016874120.1 17 - 18 3861 64
GenBank mRNA AB002296.2 - 16 - 17 3670 64

View Full Product Details