Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
Chromosome Location
Chr.5: 66596348 - 67169595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164664.1 NP_001158136.1 20 - 21 2923 84
NM_001290226.1 NP_001277155.1 18 - 19 2375 84
NM_001290227.1 NP_001277156.1 18 - 19 2147 84
NM_001297651.1 NP_001284580.1 17 - 18 2407 84
NM_015183.2 NP_055998.1 19 - 20 2116 84
XM_006714606.3 XP_006714669.1 18 - 19 2369 84
XM_006714610.2 XP_006714673.1 17 - 18 2356 84
XM_011543382.2 XP_011541684.1 22 - 23 3015 84
XM_011543384.2 XP_011541686.1 17 - 18 4074 84
XM_011543385.2 XP_011541687.1 17 - 18 5194 84
XM_011543386.2 XP_011541688.1 17 - 18 2025 84
XM_017009447.1 XP_016864936.1 23 - 24 3096 84
XM_017009448.1 XP_016864937.1 21 - 22 2979 84
XM_017009449.1 XP_016864938.1 18 - 19 2414 84
XM_017009450.1 XP_016864939.1 18 - 19 2108 84
XM_017009451.1 XP_016864940.1 18 - 19 2108 84
XM_017009452.1 XP_016864941.1 18 - 19 5423 84
XM_017009453.1 XP_016864942.1 17 - 18 12238 84
GenBank mRNA AB002301.1 - 9 - 10 1159 84
AK131421.1 - 19 - 20 2455 84
AK296707.1 - 18 - 19 2112 84
AK299538.1 - 18 - 19 2257 84
AK299706.1 - 17 - 18 2406 84
AY830839.1 - 19 - 20 2117 84

View Full Product Details