Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
-
Chromosome Location
Chr.5: 66596348 - 67169595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164664.1 NP_001158136.1 27 - 28 4086 75
NM_001290226.1 NP_001277155.1 25 - 26 3538 75
NM_001290227.1 NP_001277156.1 24 - 25 3109 75
NM_001297651.1 NP_001284580.1 24 - 25 3570 75
NM_015183.2 NP_055998.1 26 - 27 3279 75
XM_006714606.3 XP_006714669.1 25 - 26 3532 75
XM_006714610.2 XP_006714673.1 23 - 24 3318 75
XM_011543382.2 XP_011541684.1 29 - 30 4178 75
XM_011543384.2 XP_011541686.1 24 - 25 5237 75
XM_011543385.2 XP_011541687.1 24 - 25 6357 75
XM_011543386.2 XP_011541688.1 24 - 25 3188 75
XM_017009447.1 XP_016864936.1 30 - 31 4259 75
XM_017009448.1 XP_016864937.1 28 - 29 4142 75
XM_017009449.1 XP_016864938.1 25 - 26 3577 75
XM_017009450.1 XP_016864939.1 25 - 26 3271 75
XM_017009451.1 XP_016864940.1 24 - 25 3070 75
XM_017009452.1 XP_016864941.1 25 - 26 6586 75
XM_017009453.1 XP_016864942.1 24 - 25 13401 75
GenBank mRNA AB002301.1 - 16 - 17 2322 75
AY830839.1 - 26 - 27 3280 75
BX648980.1 - 6 - 7 888 75

View Full Product Details