Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 12 - 13 5314 59
NM_014866.1 NP_055681.1 13 - 14 5304 59
XM_005266134.2 XP_005266191.1 12 - 13 5167 59
XM_011519246.2 XP_011517548.1 13 - 14 5313 59
XM_011519247.1 XP_011517549.1 12 - 13 5170 59
XM_011519248.2 XP_011517550.1 12 - 13 5144 59
XM_011519249.1 XP_011517551.1 12 - 13 5167 59
XM_011519250.2 XP_011517552.1 12 - 13 5167 59
XM_011519251.1 XP_011517553.1 12 - 13 5167 59
XM_011519252.2 XP_011517554.1 12 - 13 5167 59
XM_011519253.1 XP_011517555.1 12 - 13 5167 59
XM_011519254.1 XP_011517556.1 12 - 13 5167 59
XM_011519255.2 XP_011517557.1 12 - 13 5167 59
XM_011519256.1 XP_011517558.1 12 - 13 5167 59
XM_011519257.1 XP_011517559.1 12 - 13 5167 59
XM_011519258.1 XP_011517560.1 12 - 13 5167 59
XM_011519259.1 XP_011517561.1 12 - 13 5167 59
XM_011519260.2 XP_011517562.1 12 - 13 5151 59
XM_011519261.1 XP_011517563.1 9 - 10 4720 59
XM_011519262.1 XP_011517564.1 13 - 14 4456 59
XM_011519263.2 XP_011517565.1 13 - 14 4456 59
XM_011519264.2 XP_011517566.1 12 - 13 5167 59
XM_017015363.1 XP_016870852.1 12 - 13 5167 59
GenBank mRNA AB002308.4 - 13 - 14 5304 59
AB593115.1 - 12 - 13 5314 59
BC008332.1 - 2 - 3 177 59
BC028183.1 - 8 - 9 1171 59
BC125018.1 - 11 - 12 3876 59
BC125019.1 - 12 - 13 3165 59
DQ903855.1 - 11 - 12 4461 59

View Full Product Details