The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012401.3 | NP_036533.2 | 4 - 5 | 1472 | 76 |
XM_005261909.1 | XP_005261966.1 | 5 - 6 | 1520 | 76 | |
XM_005261910.2 | XP_005261967.1 | 5 - 6 | 1531 | 76 | |
XM_005261911.4 | XP_005261968.1 | 4 - 5 | 1470 | 76 | |
XM_006724413.2 | XP_006724476.1 | 6 - 7 | 1590 | 76 | |
XM_011530682.2 | XP_011528984.1 | 5 - 6 | 1671 | 76 | |
XM_017028703.1 | XP_016884192.1 | 6 - 7 | 1571 | 76 | |
XM_017028704.1 | XP_016884193.1 | 5 - 6 | 1524 | 76 | |
GenBank mRNA | AB002313.1 | - | 3 - 4 | 1389 | 76 |
BC127770.1 | - | 5 - 6 | 1576 | 76 |