The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015347.4 | NP_056162.4 | 15 - 16 | 2987 | 66 |
XM_011538102.1 | XP_011536404.1 | 17 - 18 | 3805 | 66 | |
XM_011538103.2 | XP_011536405.1 | 19 - 20 | 4267 | 66 | |
XM_011538104.2 | XP_011536406.1 | 19 - 20 | 4082 | 66 | |
XM_011538105.2 | XP_011536407.1 | 18 - 19 | 3539 | 66 | |
XM_011538106.2 | XP_011536408.1 | 17 - 18 | 3380 | 66 | |
XM_017019105.1 | XP_016874594.1 | 19 - 20 | 4280 | 66 | |
XM_017019106.1 | XP_016874595.1 | 18 - 19 | 3539 | 66 | |
XM_017019107.1 | XP_016874596.1 | 18 - 19 | 3361 | 66 | |
XM_017019108.1 | XP_016874597.1 | 18 - 19 | 3544 | 66 | |
XM_017019109.1 | XP_016874598.1 | 16 - 17 | 3088 | 66 | |
XM_017019110.1 | XP_016874599.1 | 18 - 19 | 3361 | 66 | |
XM_017019111.1 | XP_016874600.1 | 18 - 19 | 3361 | 66 | |
GenBank mRNA | AB002316.1 | - | 15 - 16 | 2987 | 66 |
AY699979.1 | - | 12 - 13 | 2678 | 66 | |
BX641152.1 | - | 2 - 3 | 7719 | 66 |