Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 4 - 5 830 73
NM_015052.4 NP_055867.3 5 - 6 1065 73
XM_005249665.3 XP_005249722.1 4 - 5 844 73
XM_006715670.3 XP_006715733.1 3 - 4 742 73
XM_006715671.3 XP_006715734.1 3 - 4 742 73
XM_006715673.3 XP_006715736.1 3 - 4 742 73
XM_011515220.2 XP_011513522.1 4 - 5 635 73
XM_011515222.2 XP_011513524.1 3 - 4 776 73
XM_011515223.2 XP_011513525.1 3 - 4 577 73
XM_011515224.2 XP_011513526.1 3 - 4 1057 73
XM_017011882.1 XP_016867371.1 3 - 4 1084 73
XM_017011883.1 XP_016867372.1 4 - 5 1021 73
XM_017011884.1 XP_016867373.1 3 - 4 1084 73
XM_017011885.1 XP_016867374.1 5 - 6 1017 73
XM_017011886.1 XP_016867375.1 4 - 5 849 73
XM_017011887.1 XP_016867376.1 3 - 4 1084 73
XM_017011888.1 XP_016867377.1 3 - 4 1084 73
XM_017011889.1 XP_016867378.1 4 - 5 1141 73
XM_017011890.1 XP_016867379.1 4 - 5 849 73
GenBank mRNA AB002320.2 - 5 - 6 1065 73
AB048365.1 - 2 - 3 417 73
AK090799.1 - 3 - 4 1084 73
AK294918.1 - 4 - 5 784 73
BC151227.1 - 5 - 6 1065 73

View Full Product Details