Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 24 - 25 4518 100
NM_015052.4 NP_055867.3 26 - 27 4855 100
XM_005249665.3 XP_005249722.1 25 - 26 4637 100
XM_006715670.3 XP_006715733.1 24 - 25 4535 100
XM_006715671.3 XP_006715734.1 24 - 25 4532 100
XM_006715673.3 XP_006715736.1 23 - 24 4430 100
XM_011515220.2 XP_011513522.1 25 - 26 4428 100
XM_011515222.2 XP_011513524.1 24 - 25 4569 100
XM_011515223.2 XP_011513525.1 24 - 25 4370 100
XM_011515224.2 XP_011513526.1 24 - 25 4850 100
XM_011515225.2 XP_011513527.2 22 - 23 3887 100
XM_011515226.2 XP_011513528.1 21 - 22 4111 100
XM_017011882.1 XP_016867371.1 24 - 25 4877 100
XM_017011883.1 XP_016867372.1 25 - 26 4814 100
XM_017011884.1 XP_016867373.1 24 - 25 4874 100
XM_017011885.1 XP_016867374.1 26 - 27 4810 100
XM_017011886.1 XP_016867375.1 25 - 26 4639 100
XM_017011887.1 XP_016867376.1 23 - 24 4775 100
XM_017011888.1 XP_016867377.1 23 - 24 4772 100
XM_017011889.1 XP_016867378.1 24 - 25 4829 100
GenBank mRNA AB002320.2 - 26 - 27 4855 100
AB048365.1 - 23 - 24 4207 100
AK294918.1 - 24 - 25 4472 100
BC151227.1 - 26 - 27 4855 100

View Full Product Details