Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KDM6B
Entrez Gene ID
23135
Gene Name
lysine demethylase 6B
Gene Aliases
JMJD3
Chromosome Location
Chr.17: 7834210 - 7854800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080424.1 NP_001073893.1 14 - 15 4250 61
XM_005256549.3 XP_005256606.1 15 - 16 4402 61
XM_005256550.4 XP_005256607.1 14 - 15 4244 61
XM_005256551.3 XP_005256608.1 14 - 15 4283 61
XM_005256552.4 XP_005256609.1 14 - 15 5256 61
XM_006721483.3 XP_006721546.1 15 - 16 4347 61
XM_011523750.2 XP_011522052.1 15 - 16 5607 61
XM_011523752.2 XP_011522054.1 14 - 15 4247 61
XM_017024381.1 XP_016879870.1 14 - 15 5693 61
GenBank mRNA AB002344.2 - 14 - 15 4255 61
BC009994.1 - 4 - 5 879 61

View Full Product Details