Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
Chromosome Location
Chr.16: 10944488 - 11193272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243403.1 NP_001230332.1 4 - 5 724 67
NM_015226.2 NP_056041.1 4 - 5 724 67
XM_005255210.1 XP_005255267.1 4 - 5 718 67
XM_005255211.1 XP_005255268.1 4 - 5 718 67
XM_005255213.1 XP_005255270.1 4 - 5 718 67
XM_005255214.1 XP_005255271.1 4 - 5 718 67
XM_005255215.3 XP_005255272.1 4 - 5 718 67
XM_005255216.1 XP_005255273.1 4 - 5 718 67
XM_006720870.3 XP_006720933.1 4 - 5 718 67
XM_011522434.1 XP_011520736.1 4 - 5 718 67
XM_011522435.1 XP_011520737.1 4 - 5 718 67
XM_011522436.2 XP_011520738.1 4 - 5 718 67
XM_011522437.2 XP_011520739.1 4 - 5 718 67
XM_011522438.2 XP_011520740.1 4 - 5 718 67
XM_011522439.2 XP_011520741.1 4 - 5 718 67
XM_011522440.2 XP_011520742.1 4 - 5 718 67
XM_017023089.1 XP_016878578.1 4 - 5 718 67
GenBank mRNA AB002348.3 - 4 - 5 627 67
BC112897.1 - 4 - 5 631 67

View Full Product Details