Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCF2L
Entrez Gene ID
23263
Gene Name
MCF.2 cell line derived transforming sequence like
Gene Aliases
ARHGEF14, DBS, OST
Chromosome Location
Chr.13: 112894378 - 113099739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112732.2 NP_001106203.2 2 - 3 322 92
NM_001320815.1 NP_001307744.1 2 - 3 354 92
NM_001320816.1 NP_001307745.1 2 - 3 531 92
NM_001320817.1 NP_001307746.1 2 - 3 354 92
NM_024979.4 NP_079255.4 2 - 3 354 92
XM_011537482.1 XP_011535784.1 3 - 4 311 92
XM_011537483.2 XP_011535785.1 3 - 4 431 92
XM_011537484.1 XP_011535786.1 2 - 3 758 92
XM_011537485.1 XP_011535787.1 2 - 3 430 92
XM_011537486.2 XP_011535788.2 2 - 3 446 92
XM_011537487.1 XP_011535789.1 2 - 3 351 92
XM_011537488.1 XP_011535790.1 2 - 3 190 92
XM_011537490.1 XP_011535792.1 2 - 3 724 92
XM_017020493.1 XP_016875982.1 2 - 3 446 92
XM_017020494.1 XP_016875983.1 2 - 3 446 92
XM_017020495.1 XP_016875984.1 2 - 3 446 92
XM_017020497.1 XP_016875986.1 2 - 3 724 92
XM_017020499.1 XP_016875988.1 2 - 3 446 92
GenBank mRNA AB002360.1 - 2 - 3 299 92
AB116074.1 - 2 - 3 210 92
AK295047.1 - 2 - 3 351 92
AK295670.1 - 2 - 3 164 92
AK307867.1 - 2 - 3 502 92
AK315969.1 - 2 - 3 322 92
BC011853.2 - 2 - 3 314 92
BC020208.1 - 2 - 3 314 92
CR988747.1 - 2 - 3 322 92
DA114166.1 - 2 - 3 356 92

View Full Product Details