Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRUNE2
Entrez Gene ID
158471
Gene Name
prune homolog 2
Gene Aliases
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location
Chr.9: 76611376 - 76906220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308048.1 NP_001294977.1 11 - 12 8682 74
NM_001308051.1 NP_001294980.1 3 - 4 624 74
XM_005251745.1 XP_005251802.1 11 - 12 8815 74
XM_005251746.1 XP_005251803.1 11 - 12 8815 74
XM_005251750.1 XP_005251807.1 11 - 12 8815 74
XM_005251751.1 XP_005251808.1 11 - 12 8815 74
XM_005251754.1 XP_005251811.1 11 - 12 8815 74
XM_006716982.1 XP_006717045.1 11 - 12 8815 74
XM_006716984.2 XP_006717047.1 11 - 12 8815 74
XM_006716985.1 XP_006717048.1 11 - 12 8815 74
XM_011518323.1 XP_011516625.1 11 - 12 8815 74
XM_011518326.2 XP_011516628.1 11 - 12 8815 74
XM_011518327.1 XP_011516629.1 11 - 12 8815 74
XM_011518328.2 XP_011516630.1 11 - 12 8815 74
XM_017014348.1 XP_016869837.1 11 - 12 8815 74
XM_017014351.1 XP_016869840.1 11 - 12 8815 74
XM_017014353.1 XP_016869842.1 11 - 12 8815 74
XM_017014354.1 XP_016869843.1 3 - 4 413 74
XM_017014355.1 XP_016869844.1 3 - 4 413 74
XM_017014356.1 XP_016869845.1 3 - 4 413 74
XM_017014357.1 XP_016869846.1 3 - 4 413 74
XM_017014359.1 XP_016869848.1 3 - 4 413 74
GenBank mRNA AB002365.2 - 4 - 5 1751 74
AB535152.1 - 11 - 12 8555 74
AK299857.1 - 3 - 4 406 74
BC022571.1 - 3 - 4 409 74
BC150641.1 - 5 - 6 7697 74

View Full Product Details