Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUSC2
Entrez Gene ID
9853
Gene Name
RUN and SH3 domain containing 2
Gene Aliases
Iporin
Chromosome Location
Chr.9: 35487999 - 35561898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135999.1 NP_001129471.1 6 - 7 3632 95
NM_014806.3 NP_055621.1 6 - 7 3376 95
NR_052015.2 6 - 7 3051 95
XM_006716896.2 XP_006716959.1 5 - 6 3223 95
GenBank mRNA AB002373.2 - 6 - 7 3218 95
BC064843.1 - 5 - 6 2420 95
BC132766.1 - 5 - 6 3161 95
BC132770.1 - 5 - 6 3161 95
BC146654.1 - 6 - 7 3350 95
BC150211.1 - 6 - 7 2963 95
BC150262.1 - 6 - 7 3229 95

View Full Product Details