Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 2 - 3 698 101
NM_004319.2 NP_004310.1 2 - 3 698 101
NM_207108.2 NP_996991.1 2 - 3 698 101
XM_017001340.1 XP_016856829.1 2 - 3 648 101
XM_017001341.1 XP_016856830.1 2 - 3 648 101
GenBank mRNA AB006627.1 - 2 - 3 496 101
AK295126.1 - 2 - 3 650 101
AK307820.1 - 2 - 3 609 101
AK309094.1 - 2 - 3 767 101
BC142697.1 - 2 - 3 477 101
BC146795.1 - 2 - 3 496 101

View Full Product Details