Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQSEC2
Entrez Gene ID
23096
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
BRAG1, MRX1, MRX78
Chromosome Location
Chr.X: 53225784 - 53321350 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111125.2 NP_001104595.1 9 - 10 3088 94
NM_015075.1 NP_055890.1 9 - 10 2365 94
XM_006724579.2 XP_006724642.1 10 - 11 3210 94
XM_006724580.3 XP_006724643.1 9 - 10 2378 94
XM_006724581.3 XP_006724644.1 10 - 11 3210 94
XM_006724582.3 XP_006724645.1 10 - 11 3210 94
XM_006724583.3 XP_006724646.1 10 - 11 3210 94
XM_006724584.2 XP_006724647.1 10 - 11 3210 94
XM_011530773.2 XP_011529075.1 8 - 9 2282 94
XM_011530774.2 XP_011529076.1 10 - 11 3210 94
XM_011530776.1 XP_011529078.1 10 - 11 3210 94
XM_011530777.1 XP_011529079.1 10 - 11 3210 94
XM_017029359.1 XP_016884848.1 8 - 9 3084 94
XM_017029360.1 XP_016884849.1 9 - 10 2496 94
GenBank mRNA AB011094.1 - 9 - 10 3088 94
AK095232.1 - 9 - 10 2365 94

View Full Product Details