The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014825.2 | NP_055640.2 | 29 - 30 | 4950 | 101 |
GenBank mRNA | AB011111.2 | - | 24 - 25 | 4184 | 101 |
AF231919.1 | - | 29 - 30 | 4788 | 101 |