Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FNBP1
- Entrez Gene ID
-
23048
- Gene Name
- formin binding protein 1
- Gene Aliases
-
FBP17
- Chromosome Location
-
Chr.9: 129887187 - 130053846 on Build GRCh38
View Full Product Details