The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195733.1 | NP_001182662.1 | 12 - 13 | 1601 | 114 |
NM_001300849.1 | NP_001287778.1 | 12 - 13 | 1614 | 114 | |
NM_012398.2 | NP_036530.1 | 12 - 13 | 1601 | 114 | |
XM_011527845.1 | XP_011526147.1 | 12 - 13 | 1608 | 114 | |
XM_011527846.1 | XP_011526148.1 | 12 - 13 | 1608 | 114 | |
XM_011527848.1 | XP_011526150.1 | 12 - 13 | 1608 | 114 | |
XM_011527849.1 | XP_011526151.1 | 12 - 13 | 1608 | 114 | |
XM_011527850.2 | XP_011526152.1 | 12 - 13 | 1567 | 114 | |
XM_011527851.2 | XP_011526153.1 | 5 - 6 | 1195 | 114 | |
XM_011527853.2 | XP_011526155.1 | 5 - 6 | 1026 | 114 | |
XM_017026540.1 | XP_016882029.1 | 12 - 13 | 1608 | 114 | |
GenBank mRNA | AB011161.1 | - | 12 - 13 | 1569 | 114 |
AK301309.1 | - | 12 - 13 | 1584 | 114 | |
AK315912.1 | - | 12 - 13 | 1587 | 114 | |
FJ965536.1 | - | 12 - 13 | 1512 | 114 | |
FJ965537.1 | - | 12 - 13 | 1512 | 114 |