The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100913.2 | NP_001094383.2 | 15 - 16 | 2131 | 76 |
NM_001243127.2 | NP_001230056.1 | 15 - 16 | 1641 | 76 | |
NM_015197.3 | NP_056012.2 | 15 - 16 | 2119 | 76 | |
XM_006720090.1 | XP_006720153.1 | 15 - 16 | 1799 | 76 | |
XM_006720091.1 | XP_006720154.1 | 15 - 16 | 1799 | 76 | |
XM_006720092.2 | XP_006720155.1 | 15 - 16 | 1760 | 76 | |
XM_017021105.1 | XP_016876594.1 | 15 - 16 | 1787 | 76 | |
XM_017021106.1 | XP_016876595.1 | 15 - 16 | 1784 | 76 | |
XM_017021107.1 | XP_016876596.1 | 15 - 16 | 1775 | 76 | |
XM_017021108.1 | XP_016876597.1 | 15 - 16 | 1772 | 76 | |
XM_017021109.1 | XP_016876598.1 | 15 - 16 | 1796 | 76 | |
XM_017021110.1 | XP_016876599.1 | 15 - 16 | 1787 | 76 | |
XM_017021111.1 | XP_016876600.1 | 15 - 16 | 1665 | 76 | |
XM_017021112.1 | XP_016876601.1 | 15 - 16 | 1567 | 76 | |
GenBank mRNA | AB011174.1 | - | 15 - 16 | 1802 | 76 |
AY320284.2 | - | 15 - 16 | 2118 | 76 | |
BC010663.1 | - | 12 - 13 | 1226 | 76 | |
BC050351.1 | - | 15 - 16 | 1657 | 76 | |
BC065220.1 | - | 15 - 16 | 1732 | 76 | |
BC131591.1 | - | 15 - 16 | 1619 | 76 |