The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015188.1 | NP_056003.1 | 1 - 2 | 1152 | 84 |
XM_006717732.3 | XP_006717795.2 | 1 - 2 | 1152 | 84 | |
XM_006717733.2 | XP_006717796.1 | 1 - 2 | 76 | 84 | |
XM_017015996.1 | XP_016871485.1 | 1 - 2 | 1152 | 84 | |
XM_017015997.1 | XP_016871486.1 | 1 - 2 | 1152 | 84 | |
GenBank mRNA | AB011180.1 | - | 1 - 2 | 968 | 84 |
AB449898.1 | - | 1 - 2 | 949 | 84 |