The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015188.1 | NP_056003.1 | 2 - 3 | 1285 | 98 |
XM_006717732.3 | XP_006717795.2 | 2 - 3 | 1285 | 98 | |
XM_011539558.2 | XP_011537860.1 | 2 - 3 | 296 | 98 | |
XM_017015996.1 | XP_016871485.1 | 2 - 3 | 1285 | 98 | |
XM_017015997.1 | XP_016871486.1 | 2 - 3 | 1285 | 98 | |
GenBank mRNA | AB011180.1 | - | 2 - 3 | 1101 | 98 |
AB449898.1 | - | 2 - 3 | 1082 | 98 |