The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080497.2 | NP_001073966.2 | 2 - 3 | 932 | 68 |
GenBank mRNA | AB011542.1 | - | 1 - 2 | 124 | 68 |
AK295380.1 | - | 2 - 3 | 932 | 68 |