Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
Chromosome Location
Chr.7: 51016212 - 51316818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287436.1 NP_001274365.1 10 - 11 1860 67
NM_015198.3 NP_056013.2 9 - 10 1689 67
XM_005271750.1 XP_005271807.1 11 - 12 1896 67
XM_005271751.1 XP_005271808.1 10 - 11 1851 67
XM_005271756.1 XP_005271813.1 9 - 10 1680 67
XM_011515234.1 XP_011513536.1 12 - 13 1971 67
XM_011515235.1 XP_011513537.1 11 - 12 1926 67
XM_011515236.1 XP_011513538.1 12 - 13 1971 67
XM_011515237.1 XP_011513539.1 11 - 12 1800 67
XM_011515238.1 XP_011513540.1 12 - 13 1971 67
XM_011515239.1 XP_011513541.1 11 - 12 1800 67
XM_011515240.1 XP_011513542.1 10 - 11 1755 67
XM_011515241.1 XP_011513543.1 6 - 7 975 67
XM_017011898.1 XP_016867387.1 10 - 11 1725 67
XM_017011899.1 XP_016867388.1 10 - 11 1851 67
XM_017011900.1 XP_016867389.1 9 - 10 1680 67
GenBank mRNA AB014533.1 - 9 - 10 1670 67
AK293883.1 - 2 - 3 226 67
AL713786.1 - 1 - 2 5144 67
BC029275.1 - 4 - 5 560 67
BC045771.1 - 8 - 9 1159 67
BC136441.1 - 10 - 11 1775 67
BC144099.1 - 9 - 10 1604 67
BC150263.1 - 9 - 10 1670 67

View Full Product Details