The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014010.4 | NP_054729.3 | 13 - 14 | 2469 | 119 |
GenBank mRNA | AB014534.1 | - | 13 - 14 | 2469 | 119 |
AF116574.1 | - | 11 - 12 | 1690 | 119 | |
AK127350.1 | - | 12 - 13 | 1690 | 119 | |
AK315025.1 | - | 11 - 12 | 1533 | 119 | |
BC018759.2 | - | 7 - 8 | 918 | 119 | |
BC146756.1 | - | 13 - 14 | 2469 | 119 | |
BX649176.1 | - | 13 - 14 | 1804 | 119 |