The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103146.1 | NP_001096616.1 | 25 - 26 | 3498 | 80 |
NM_001103147.1 | NP_001096617.1 | 27 - 28 | 3631 | 80 | |
NM_001103148.1 | NP_001096618.1 | 24 - 25 | 3414 | 80 | |
NM_015575.3 | NP_056390.2 | 27 - 28 | 3638 | 80 | |
GenBank mRNA | AB014542.2 | - | 27 - 28 | 3590 | 80 |
AK027021.1 | - | 4 - 5 | 783 | 80 | |
AY176045.1 | - | 27 - 28 | 3612 | 80 | |
BC008072.2 | - | 1 - 2 | 142 | 80 | |
BC136251.1 | - | 24 - 25 | 3369 | 80 | |
BC146775.1 | - | 27 - 28 | 3590 | 80 | |
BX537885.1 | - | 25 - 26 | 3433 | 80 | |
BX538172.1 | - | 24 - 25 | 3382 | 80 | |
BX538321.1 | - | 25 - 26 | 3454 | 80 | |
BX647172.1 | - | 9 - 10 | 2336 | 80 |